Learn More
Medchemexpress LLC JMV 2959 hydrochloride | 2448414-54-6 | 545.08 | 10 MG
SDP
Catalog No. 5000447952
Encompass_Preferred
JMV 2959 hydrochloride is a potent growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist. It demonstrates an IC50 of 32±3 nM in LLC-PK1 cells and a dissociation constant (Kb) of 19±6 nM. This compound does not induce intracellular calcium mobilization on its own.
  • Antagonist of growth hormone secretagogue receptor type 1a (GHS-R1a)
  • IC50 of 32±3 nM in LLC-PK1 cells
  • Dissociation constant (Kb) of 19±6 nM
  • Does not induce intracellular calcium mobilization
  • Attenuates phencyclidine (PCP)-induced deficits in prepulse inhibition in vivo
Catalog No. 50-004-47952 Supplier Medchemexpress LLC Supplier No. HYU00433A10MG
JMV 2959 hydrochloride is a potent growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist. It demonstrates an IC50 of 32±3 nM in LLC-PK1 cells and a dissociation constant (Kb) of 19±6 nM. This compound does not induce intracellular calcium mobilization on its own.
  • Antagonist of growth hormone secretagogue receptor type 1a (GHS-R1a)
  • IC50 of 32±3 nM in LLC-PK1 cells
  • Dissociation constant (Kb) of 19±6 nM
  • Does not induce intracellular calcium mobilization
  • Attenuates phencyclidine (PCP)-induced deficits in prepulse inhibition in vivo