Learn More
Medchemexpress LLC JMV 2959 hydrochloride | 2448414-54-6 | 545.08 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYU00433A10MM/1ML
Encompass_Preferred
JMV 2959 hydrochloride is a potent growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32±3 nM in LLC-PK1 cells. It does not induce intracellular calcium mobilization. Studies show it dose-dependently decreases acoustic startle responses (ASR) and increases prepulse inhibition (PPI), also blocking Phencyclidine (PCP)-induced deficits.
  • GHSR-1a antagonist (IC50 of 32±3 nM)
  • No intracellular calcium mobilization
  • Dissociation constant (Kb) of 19±6 nM
  • Decreases acoustic startle responses (ASR) and increases prepulse inhibition (PPI)
  • Blocks PCP-induced PPI deficits
  • Relevant for neurological disease and endocrinology research
  • Store at 4°C (sealed, away from moisture) or in solvent at -80°C (6 months) / -20°C (1 month)
Catalog No. 50-004-49638