Learn More
TARGETMOL CHEMICALS INC JMV 2959 hydrochloride 925238-
SDP
Catalog No. 502880610
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JMV 2959 hydrochloride (925238-89-7 free base) is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32±3 nM in LLC-PK1 cells. Purity 98.09%
Catalog No. 50-288-0610 Supplier TARGETMOL CHEMICALS INC Supplier No. T1171910MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JMV 2959 hydrochloride (925238-89-7 free base) is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32±3 nM in LLC-PK1 cells. Purity 98.09%