Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JNJ-1013 | 2597343-08-1 | 99.8% | 878.05 g/mol | C46H55N9O7S | 10 MG
SDP

Catalog No. 5000285749
Encompass_Preferred

JNJ-1013 is a research-grade PROTAC degrader that selectively targets interleukin-1 receptor-associated kinase 1 (IRAK1). It induces IRAK1 degradation, modulates downstream signaling, and has been characterized in cellular assays for apoptosis and pathway inhibition. Supplied as a high-purity solid, it is intended for in vitro biochemical and cellular studies.

  • Potent IRAK1 activity with reported IC50 values: 72 nM (IRAK1), 443 nM (IRAK4), 1,071 nM (VHL FP).
  • High reported purity (99.8%).
  • Molecular weight 878.05 g/mol; formula C46H55N9O7S.
  • Soluble in DMSO at 25 mg/mL; may require warming or sonication.
  • Recommended storage: protect from light at 4°C; in solution store at -80°C for long-term or -20°C up to one month.
  • Suitable for in vitro target validation, signaling pathway studies, and apoptosis assays.

Catalog No. 50-002-85749 Supplier Medchemexpress LLC Supplier No. HY15318810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JNJ-1013 is a research-grade PROTAC degrader that selectively targets interleukin-1 receptor-associated kinase 1 (IRAK1). It induces IRAK1 degradation, modulates downstream signaling, and has been characterized in cellular assays for apoptosis and pathway inhibition. Supplied as a high-purity solid, it is intended for in vitro biochemical and cellular studies.

  • Potent IRAK1 activity with reported IC50 values: 72 nM (IRAK1), 443 nM (IRAK4), 1,071 nM (VHL FP).
  • High reported purity (99.8%).
  • Molecular weight 878.05 g/mol; formula C46H55N9O7S.
  • Soluble in DMSO at 25 mg/mL; may require warming or sonication.
  • Recommended storage: protect from light at 4°C; in solution store at -80°C for long-term or -20°C up to one month.
  • Suitable for in vitro target validation, signaling pathway studies, and apoptosis assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.