Learn More
Medchemexpress LLC JNJ-1013 | 2597343-08-1 | C46H55N9O7S | 100 MG
SDP
Catalog No. 5000373520
Encompass_Preferred
JNJ-1013 is a potent and selective IRAK1 PROTAC degrader. It functions by inducing apoptosis and increasing the expression of cleaved PARP. This compound also plays a role in decreasing the expression of IRAK1, p-IKBα, and pSTAT3(Tyr705).
  • Potent and selective IRAK1 PROTAC degrader
  • Induces apoptosis
  • Increases the expression of cleaved PARP
  • Decreases the expression of IRAK1, p-IKBα, and pSTAT3(Tyr705)
Catalog No. 50-003-73520 Supplier Medchemexpress LLC Supplier No. HY153188100MG
JNJ-1013 is a potent and selective IRAK1 PROTAC degrader. It functions by inducing apoptosis and increasing the expression of cleaved PARP. This compound also plays a role in decreasing the expression of IRAK1, p-IKBα, and pSTAT3(Tyr705).
  • Potent and selective IRAK1 PROTAC degrader
  • Induces apoptosis
  • Increases the expression of cleaved PARP
  • Decreases the expression of IRAK1, p-IKBα, and pSTAT3(Tyr705)