Learn More
Medchemexpress LLC JNJ-1013 | 2597343-08-1 | C46H55N9O7S | 50 MG
SDP
Catalog No. 5000372529
Encompass_Preferred
JNJ-1013 is a potent and selective IRAK1 PROTAC degrader. It induces apoptosis, increases the expression of cleaved PARP, and decreases the expression of IRAK1, p-IKBα, and pSTAT3(Tyr705).
  • Potent and selective IRAK1 PROTAC degrader
  • IC50s of 72 nM for IRAK1, 443 nM for IRAK4, and 1071 nM for VHL FP
  • Induces apoptosis and increases cleaved PARP expression
  • Decreases expression of IRAK1, p-IKBα, and pSTAT3(Tyr705)
Catalog No. 50-003-72529 Supplier Medchemexpress LLC Supplier No. HY15318850MG
JNJ-1013 is a potent and selective IRAK1 PROTAC degrader. It induces apoptosis, increases the expression of cleaved PARP, and decreases the expression of IRAK1, p-IKBα, and pSTAT3(Tyr705).
  • Potent and selective IRAK1 PROTAC degrader
  • IC50s of 72 nM for IRAK1, 443 nM for IRAK4, and 1071 nM for VHL FP
  • Induces apoptosis and increases cleaved PARP expression
  • Decreases expression of IRAK1, p-IKBα, and pSTAT3(Tyr705)