Learn More
Cayman Chemical JNJ-10181457 (hydrochloride), Purity: ≥98%, CAS: 544707-20-2, 1 mg
SDP
Catalog No. 502954891
Encompass_Preferred
A histamine H3 receptor antagonist (Kis = 1.17 and 7.08 nM for human and rat receptors, respectively); increases extracellular norepinephrine and acetylcholine levels in the frontal cortex of rats at 10 mg/kg; reduces the number of cataplectic attacks and time spent in cataplexy in familial narcoleptic Dobermans at 1.25-10 mg/kg; decreases the time spent in the open areas of the elevated zero maze in mice at 10 mg/kg, as well as increases locomotor activity in an open field test; inhibits LPS-induced increases in time spent immobile in the tail suspension test in mice.
Catalog No. 50-295-4891 Supplier Cayman Chemical Supplier No. 119971 MG
A histamine H3 receptor antagonist (Kis = 1.17 and 7.08 nM for human and rat receptors, respectively); increases extracellular norepinephrine and acetylcholine levels in the frontal cortex of rats at 10 mg/kg; reduces the number of cataplectic attacks and time spent in cataplexy in familial narcoleptic Dobermans at 1.25-10 mg/kg; decreases the time spent in the open areas of the elevated zero maze in mice at 10 mg/kg, as well as increases locomotor activity in an open field test; inhibits LPS-induced increases in time spent immobile in the tail suspension test in mice.