Learn More
Medchemexpress LLC HY-W011266 1mg Medchemexpress, JNJ-​10198409 CAS:627518-40-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0112661MG
Encompass_Preferred
Medchemexpress, HY-W011266 1mg JNJ-​10198409 CAS:627518-40-5 JNJ-10198409 is a relatively selective, orally active, and ATP competitive PDGF-RTK (platelet-derived growth factor receptor tyrosine kinase) inhibitor (IC50=2 nM). It is a dual-mechanism, antiangiogenic, and tumor cell antiproliferative agent. JNJ-10198409 has good activity against PDGFR-β kinase (IC50=4.2 nM) and PDGFR-α kinase (IC50=45 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0681