Learn More
Medchemexpress LLC HY-19889 5mg Medchemexpress, JNJ-18038683 CAS:851376-05-1 Purity:>98%
SDP
Catalog No. 501970042
Encompass_Preferred
Medchemexpress, HY-19889 5mg JNJ-18038683 CAS:851376-05-1 JNJ-18038683 is a 5-Hydroxytryptamine Type 7 (5-HT7) receptor antagonist, with pKis of 8.19, 8.20 for rat and human 5-HT7 in HEK293 cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0042 Supplier Medchemexpress LLC Supplier No. HY198895MG
Medchemexpress, HY-19889 5mg JNJ-18038683 CAS:851376-05-1 JNJ-18038683 is a 5-Hydroxytryptamine Type 7 (5-HT7) receptor antagonist, with pKis of 8.19, 8.20 for rat and human 5-HT7 in HEK293 cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.