Learn More
Medchemexpress LLC JNJ-18038683 | 851376-05-1 | 98.3% | 529.97 | 100 MG
SDP
Catalog No. 5000400461
Encompass_Preferred
JNJ-18038683 is a 5-Hydroxytryptamine Type 7 (5-HT7) receptor antagonist.
  • High affinity for rat and human 5-HT7 receptors.
  • Suitable for research use only.
  • Suppresses REM sleep in vivo.
Catalog No. 50-004-00461 Supplier Medchemexpress LLC Supplier No. HY19889100MG
JNJ-18038683 is a 5-Hydroxytryptamine Type 7 (5-HT7) receptor antagonist.
  • High affinity for rat and human 5-HT7 receptors.
  • Suitable for research use only.
  • Suppresses REM sleep in vivo.