Learn More
Medchemexpress LLC JNJ-18038683 | 851376-05-1 | 98.31% | 529.97 | 50 MG
SDP
Catalog No. 5000402023
Encompass_Preferred
JNJ-18038683 is a 5-Hydroxytryptamine Type 7 (5-HT7) receptor antagonist, with pKis of 8.19 and 8.20 for rat and human 5-HT7 in HEK293 cells. It displaces specific [3H]5-CT binding sites from rat and human 5-HT7 receptor expressed in HEK293 cells with high affinity. In vivo, it dose-dependently suppresses REM sleep.
  • High affinity for 5-HT7 receptors (rat and human)
  • Antagonizes 5-HT (100 nM)-stimulated adenylyl cyclase activity
  • Dose-dependently suppresses REM sleep
  • Maintains decreased REM sleep time with repeated administration
Catalog No. 50-004-02023 Supplier Medchemexpress LLC Supplier No. HY1988950MG
JNJ-18038683 is a 5-Hydroxytryptamine Type 7 (5-HT7) receptor antagonist, with pKis of 8.19 and 8.20 for rat and human 5-HT7 in HEK293 cells. It displaces specific [3H]5-CT binding sites from rat and human 5-HT7 receptor expressed in HEK293 cells with high affinity. In vivo, it dose-dependently suppresses REM sleep.
  • High affinity for 5-HT7 receptors (rat and human)
  • Antagonizes 5-HT (100 nM)-stimulated adenylyl cyclase activity
  • Dose-dependently suppresses REM sleep
  • Maintains decreased REM sleep time with repeated administration