Learn More
Apexbio Technology LLC JNJ-26481585, 10mg. Cas: 875320-29-9 MFCD: MFCD17010272. Potent HDAC inhibitor
SDP
Catalog No. 501904875
Encompass_Preferred
JNJ-26481585 is a novel histone deacetylase (HDAC) inhibitor that exerts a strong potency towards class I HDAC (including HDAC1, HDAC2 and HDAC3), with values of inhibition constant IC50 of 0.11, 0.33 and 4.8 nmol/L respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4875 Supplier Apexbio Technology LLC Supplier No. A409010
JNJ-26481585 is a novel histone deacetylase (HDAC) inhibitor that exerts a strong potency towards class I HDAC (including HDAC1, HDAC2 and HDAC3), with values of inhibition constant IC50 of 0.11, 0.33 and 4.8 nmol/L respectively. Other sizes are available. Please inqury us for quote.