Learn More
Apexbio Technology LLC JNJ-26481585 875320-29-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063973
Encompass_Preferred
JNJ-26481585 (CAS 875320-29-9) is a small-molecule inhibitor targeting class I histone deacetylases (HDACs) specifically HDAC1 HDAC2 and HDAC3 with reported IC50 values of 0 11 0 33 and 4 8 nM respectively It exhibits potent anti-proliferative activity across various human cancer cell lines including lung breast colon prostate ovarian and brain cancers (IC50 range 3 1 246 nM) Mechanistically JNJ-26481585 induces histone H3 acetylation alleviates HDAC-mediated repression of the p21waf1 cip1 promoter and promotes apoptosis in colorectal cancer cell models Consequently this compound serves as a research tool for cancer epigenetics and therapeutic assessment
Catalog No. 50-000-63973 Supplier Apexbio Technology LLC Supplier No. A40905.1
JNJ-26481585 (CAS 875320-29-9) is a small-molecule inhibitor targeting class I histone deacetylases (HDACs) specifically HDAC1 HDAC2 and HDAC3 with reported IC50 values of 0 11 0 33 and 4 8 nM respectively It exhibits potent anti-proliferative activity across various human cancer cell lines including lung breast colon prostate ovarian and brain cancers (IC50 range 3 1 246 nM) Mechanistically JNJ-26481585 induces histone H3 acetylation alleviates HDAC-mediated repression of the p21waf1 cip1 promoter and promotes apoptosis in colorectal cancer cell models Consequently this compound serves as a research tool for cancer epigenetics and therapeutic assessment