Learn More
Cayman Chemical JNJ-26854165, Purity: ≥98%, CAS: 881202-45-5, 5 mg
SDP
Catalog No. 502972256
Encompass_Preferred
An antagonist of MDM2 that suppresses the growth of cancer cell lines expressing wild-type p53 (IC50 values range from 240-440 nM); orally bioavailable, having anti-proliferation as well as apoptotic actions in various tumor models; affects multiple signaling pathways
Catalog No. 50-297-2256 Supplier Cayman Chemical Supplier No. 162595 MG
An antagonist of MDM2 that suppresses the growth of cancer cell lines expressing wild-type p53 (IC50 values range from 240-440 nM); orally bioavailable, having anti-proliferation as well as apoptotic actions in various tumor models; affects multiple signaling pathways