Learn More
Selleck Chemical LLC JNJ-26854165 (Serdemetan) 5mg 881202-45-5
SDP
Catalog No. 508286
Encompass_Preferred
JNJ-26854165 (Serdemetan) acts as a HDM2 ubiquitin ligase antagonist and also induces early apoptosis in p53 wild-type cells, inhibits cellular proliferation followed by delayed apoptosis in the absence of functional p53. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-828-6 Supplier Selleck Chemical LLC Supplier No. S11725MG
Add to Cart
Add to Cart
JNJ-26854165 (Serdemetan) acts as a HDM2 ubiquitin ligase antagonist and also induces early apoptosis in p53 wild-type cells, inhibits cellular proliferation followed by delayed apoptosis in the absence of functional p53. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.