Learn More
Selleck Chemical LLC JNJ-26854165 (Serdemetan) 25mg 881202-45-5
SDP
Catalog No. 508287
Encompass_Preferred
JNJ-26854165 (Serdemetan) acts as a HDM2 ubiquitin ligase antagonist and also induces early apoptosis in p53 wild-type cells, inhibits cellular proliferation followed by delayed apoptosis in the absence of functional p53. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-828-7 Supplier Selleck Chemical LLC Supplier No. S117225MG
JNJ-26854165 (Serdemetan) acts as a HDM2 ubiquitin ligase antagonist and also induces early apoptosis in p53 wild-type cells, inhibits cellular proliferation followed by delayed apoptosis in the absence of functional p53. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.