Learn More
Medchemexpress LLC JNJ 303 | 878489-28-2 | 98.7% | 440.98 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1695310MM/1ML
Encompass_Preferred
JNJ-303 is a specific delayed rectifier Kv blocker that potently blocks IKs with an IC50 value of 64 nM. It can be used for the research of diabetes, obesity, and the central nervous system.
  • Specific delayed rectifier Kv blocker.
  • Potently blocks IKs with an IC50 value of 64 nM.
  • Useful for research related to diabetes, obesity, and the central nervous system.
  • Recapitulates the Exn4-induced decrease in fasting blood glucose level in mice.
Catalog No. 50-003-90535