Learn More
Medchemexpress LLC JNJ 303 | 878489-28-2 | 98.7% | 440.98 | 100 MG
SDP
Catalog No. 5000391275
Encompass_Preferred
JNJ-303 is a specific delayed rectifier Kv blocker that can potently block IKs with an IC50 value of 64 nM. It can be used for research on diabetes, obesity, and the central nervous system.
  • Specific delayed rectifier Kv blocker.
  • Potent IKs blocker with an IC50 value of 64 nM.
  • Can be used for research of diabetes, obesity, and central nervous system.
  • In vitro studies: does not affect other cardiac channels but induces QT-prolongations and causes unprovoked torsades de pointes (TdP).
  • In vivo studies: recapitulates the Exn4-induced decrease in fasting blood glucose level in mice.
Catalog No. 50-003-91275 Supplier Medchemexpress LLC Supplier No. HY16953100MG
JNJ-303 is a specific delayed rectifier Kv blocker that can potently block IKs with an IC50 value of 64 nM. It can be used for research on diabetes, obesity, and the central nervous system.
  • Specific delayed rectifier Kv blocker.
  • Potent IKs blocker with an IC50 value of 64 nM.
  • Can be used for research of diabetes, obesity, and central nervous system.
  • In vitro studies: does not affect other cardiac channels but induces QT-prolongations and causes unprovoked torsades de pointes (TdP).
  • In vivo studies: recapitulates the Exn4-induced decrease in fasting blood glucose level in mice.