Learn More
Medchemexpress LLC JNJ 303 | 878489-28-2 | 98.74% | 440.98 | 50 MG
SDP
Catalog No. 5000391222
Encompass_Preferred
JNJ-303 is a specific delayed rectifier Kv blocker. JNJ 303 can potent block IKs with an IC50 value of 64 nM. JNJ-303 can be used for the research of diabetes, obesity and central nervous system.
  • IC50 & Target: IC50: 64 nM (IKs)
  • In Vitro:
    • Can block IKs with an IC50 value of 64 nM.
    • Does not have any effects on other cardiac channels.
    • Induces QT-prolongations and causes unprovoked torsades de pointes (TdP).
  • In Vivo: Recapitulates the Exn4-induced decrease in fasting blood glucose level in mice.
Catalog No. 50-003-91222 Supplier Medchemexpress LLC Supplier No. HY1695350MG
JNJ-303 is a specific delayed rectifier Kv blocker. JNJ 303 can potent block IKs with an IC50 value of 64 nM. JNJ-303 can be used for the research of diabetes, obesity and central nervous system.
  • IC50 & Target: IC50: 64 nM (IKs)
  • In Vitro:
    • Can block IKs with an IC50 value of 64 nM.
    • Does not have any effects on other cardiac channels.
    • Induces QT-prolongations and causes unprovoked torsades de pointes (TdP).
  • In Vivo: Recapitulates the Exn4-induced decrease in fasting blood glucose level in mice.