Learn More
TARGETMOL CHEMICALS INC JNJ-31020028 10MG
SDP
Catalog No. 502577252
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. JNJ-31020028 is a selective brain penetrant antagonist of neuropeptide Y2 receptor with high affinity(pIC50=8.07 human; pIC50=8.22 rat); it exhibits >100-fold selectivity for Y2 versus human Y1/Y4/Y5 receptors. purity: 96%
Catalog No. 50-257-7252 Supplier TARGETMOL CHEMICALS INC Supplier No. T199710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. JNJ-31020028 is a selective brain penetrant antagonist of neuropeptide Y2 receptor with high affinity(pIC50=8.07 human; pIC50=8.22 rat); it exhibits >100-fold selectivity for Y2 versus human Y1/Y4/Y5 receptors. purity: 96%