Learn More
Selleck Chemical LLC JNJ 31020028
SDP
Catalog No. 5000002956
Encompass_Preferred
JNJ 31020028 is a novel neuropeptide Y Y2 receptor antagonist which bound with high affinity with human and rat Y2 receptor (pIC50 8 07 0 05 for human Y2 receptor and pIC50 8 22 0 06 for rat Y2 receptor) and 100-fold selective versus human Y1 Y4 and Y5 receptors
Catalog No. 50-000-02956 Supplier Selleck Chemical LLC Supplier No. S64165MG
JNJ 31020028 is a novel neuropeptide Y Y2 receptor antagonist which bound with high affinity with human and rat Y2 receptor (pIC50 8 07 0 05 for human Y2 receptor and pIC50 8 22 0 06 for rat Y2 receptor) and 100-fold selective versus human Y1 Y4 and Y5 receptors