Learn More
Medchemexpress LLC JNJ-37822681 dihydrochloride | 2108806-02-4 | 98.7% | 445.26 | 100 MG
SDP
Catalog No. 5000343911
Encompass_Preferred
JNJ-37822681 dihydrochloride is a potent, specific, centrally active, fast-dissociating dopamine D2 receptor antagonist with a moderate binding affinity for the dopamine D2L receptor (Ki =158 nM), which has potential for the treatment of schizophrenia and bipolar disorder.
  • Potent and specific D2 receptor antagonist
  • Centrally active
  • Fast-dissociating
  • Moderate binding affinity for D2L receptor (Ki = 158 nM)
  • Potential for treating schizophrenia and bipolar disorder
Catalog No. 50-003-43911 Supplier Medchemexpress LLC Supplier No. HY111066A50MG
JNJ-37822681 dihydrochloride is a potent, specific, centrally active, fast-dissociating dopamine D2 receptor antagonist with a moderate binding affinity for the dopamine D2L receptor (Ki =158 nM), which has potential for the treatment of schizophrenia and bipolar disorder.
  • Potent and specific D2 receptor antagonist
  • Centrally active
  • Fast-dissociating
  • Moderate binding affinity for D2L receptor (Ki = 158 nM)
  • Potential for treating schizophrenia and bipolar disorder