Learn More
Cayman Chemical JNJ-38877605, Purity: ≥98%, CAS: 943540-75-8, 5 mg
SDP
Catalog No. 502939171
Encompass_Preferred
An ATP-competitive inhibitor of Met kinase (IC50 = 4 nM); at least 600-fold selective for Met in a panel of ~250 tyrosine and serine-threonine kinases; inhibits the growth of cancer cell lines with MET gene amplification (IC50s = 11-50 nM) but has no effect on the growth of control cells with normal MET gene copy number or no MET expression; sensitizes tumors to radiotherapy in U251 glioma and MDA-MB-231 breast cancer mouse xenograft models and increases apoptosis in irradiated tumors in the MDA-MB-231 mouse xenograft model at 50 mg/kgp; reduces tumor size by 6-fold and the number of blood vessels in tumors by 80% in an RU-P melanoma mouse xenograft model at 20 mg/kg
Catalog No. 50-293-9171 Supplier Cayman Chemical Supplier No. 276505 MG
An ATP-competitive inhibitor of Met kinase (IC50 = 4 nM); at least 600-fold selective for Met in a panel of ~250 tyrosine and serine-threonine kinases; inhibits the growth of cancer cell lines with MET gene amplification (IC50s = 11-50 nM) but has no effect on the growth of control cells with normal MET gene copy number or no MET expression; sensitizes tumors to radiotherapy in U251 glioma and MDA-MB-231 breast cancer mouse xenograft models and increases apoptosis in irradiated tumors in the MDA-MB-231 mouse xenograft model at 50 mg/kgp; reduces tumor size by 6-fold and the number of blood vessels in tumors by 80% in an RU-P melanoma mouse xenograft model at 20 mg/kg