Learn More
Selleck Chemical LLC JNJ-38877605 5mg 943540-75-8
SDP
Catalog No. 507992
Encompass_Preferred
JNJ-38877605 is an ATP-competitive inhibitor of c-Met with IC50 of 4 nM, 600-fold selective for c-Met than 200 other tyrosine and serine-threonine kinases. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-799-2 Supplier Selleck Chemical LLC Supplier No. S11145MG
JNJ-38877605 is an ATP-competitive inhibitor of c-Met with IC50 of 4 nM, 600-fold selective for c-Met than 200 other tyrosine and serine-threonine kinases. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.