Learn More
Medchemexpress LLC HY-111050 5mg Medchemexpress, JNJ-38877618 CAS:943540-74-7 Purity:>98%
SDP
Catalog No. 501968815
Encompass_Preferred
Medchemexpress, HY-111050 5mg JNJ-38877618 CAS:943540-74-7 JNJ-38877618 is a potent, highly selective, orally bioavailable Met kinase inhibitor with IC50s of 2 and 3 nM for wild type and mutant Met, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8815 Supplier Medchemexpress LLC Supplier No. HY1110505MG
Medchemexpress, HY-111050 5mg JNJ-38877618 CAS:943540-74-7 JNJ-38877618 is a potent, highly selective, orally bioavailable Met kinase inhibitor with IC50s of 2 and 3 nM for wild type and mutant Met, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.