Learn More
TARGETMOL CHEMICALS INC JNJ-39393406 5MG
SDP
Catalog No. 502580587
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. JNJ-39393406 is an (alpha)7 nicotinic acetylcholine receptor positive allosteric modulator for the study of tobacco dependence and Alzheimer's disease. purity: 98%
Catalog No. 50-258-0587 Supplier TARGETMOL CHEMICALS INC Supplier No. T276795MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. JNJ-39393406 is an (alpha)7 nicotinic acetylcholine receptor positive allosteric modulator for the study of tobacco dependence and Alzheimer's disease. purity: 98%