Learn More
Medchemexpress LLC HY-101189 5mg Medchemexpress, JNJ-39758979 CAS:1046447-90-8 Purity:>98%
SDP
Catalog No. 501932063
Encompass_Preferred
Medchemexpress, HY-101189 5mg JNJ-39758979 CAS:1046447-90-8 JNJ-39758979 is a selective, high-affinity histamine H 4 receptor antagonist with a K i of 12.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2063 Supplier Medchemexpress LLC Supplier No. HY1011895MG
Medchemexpress, HY-101189 5mg JNJ-39758979 CAS:1046447-90-8 JNJ-39758979 is a selective, high-affinity histamine H 4 receptor antagonist with a K i of 12.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.