Learn More
Cayman Chemical JNJ-42153605, Purity: ≥98%, CAS: 1254977-87-1, 5 mg
SDP
Catalog No. 502987595
Encompass_Preferred
A positive allosteric modulator of mGlurR2 (EC50 = 17 nM in CHO cells expressing the human receptor); inhibits mGluR2-mediated REM sleep in rats at 3 mg/kg; reverses phencyclidine-induced hyperlocomotion in mice (ED50 = 5.4 mg/kg)
Catalog No. 50-298-7595 Supplier Cayman Chemical Supplier No. 219845 MG
A positive allosteric modulator of mGlurR2 (EC50 = 17 nM in CHO cells expressing the human receptor); inhibits mGluR2-mediated REM sleep in rats at 3 mg/kg; reverses phencyclidine-induced hyperlocomotion in mice (ED50 = 5.4 mg/kg)