Learn More
Medchemexpress LLC HY-18162 5mg Medchemexpress, JNJ-42153605 CAS:1254977-87-1 Purity:>98%
SDP
Catalog No. 502082021
Encompass_Preferred
Medchemexpress, HY-18162 5mg JNJ-42153605 CAS:1254977-87-1 JNJ-42153605 is a positive allosteric modulator of the metabotropic glutamate 2 (mGlu2) receptor with an EC50 of 17 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2021 Supplier Medchemexpress LLC Supplier No. HY181625MG
Medchemexpress, HY-18162 5mg JNJ-42153605 CAS:1254977-87-1 JNJ-42153605 is a positive allosteric modulator of the metabotropic glutamate 2 (mGlu2) receptor with an EC50 of 17 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.