Learn More
Medchemexpress LLC JNJ-42253432 | 1428327-35-8 | 98.2% | 446.63 g/mol | C28H38N4O | 50MG
SDP
Catalog No. 5000236699
Encompass_Preferred
JNJ-42253432 is a CNS-penetrant, high-affinity, orally active P2X7 receptor antagonist used for preclinical research into P2X7-mediated signaling and neuroinflammatory processes. It is supplied as a solid with high reported purity and storage recommendations for both powder and solution forms.
  • CNS-penetrant P2X7 receptor antagonist suitable for in vitro and in vivo studies.
  • High affinity with reported pKi values of 9.1 for rat and 7.9 for human receptors.
  • High purity (98.2%).
  • White to off-white solid appearance.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-36699 Supplier Medchemexpress LLC Supplier No. HY12348150MG
JNJ-42253432 is a CNS-penetrant, high-affinity, orally active P2X7 receptor antagonist used for preclinical research into P2X7-mediated signaling and neuroinflammatory processes. It is supplied as a solid with high reported purity and storage recommendations for both powder and solution forms.
  • CNS-penetrant P2X7 receptor antagonist suitable for in vitro and in vivo studies.
  • High affinity with reported pKi values of 9.1 for rat and 7.9 for human receptors.
  • High purity (98.2%).
  • White to off-white solid appearance.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).