Learn More
Medchemexpress LLC JNJ-42253432 | 1428327-35-8 | MFCD34676648 | 98.2% | 446.63 g/mol | C28H38N4O | 100MG
SDP
Catalog No. 5000235607
Encompass_Preferred
JNJ-42253432 is a CNS-penetrant, orally active antagonist of the P2X7 purinergic receptor used in preclinical research to probe P2X7-mediated signaling and inflammatory pathways. It is supplied as a high-purity research compound and is commonly used in both in vitro and in vivo pharmacological studies.
Catalog No. 50-002-35607 Supplier Medchemexpress LLC Supplier No. HY123481100MG
JNJ-42253432 is a CNS-penetrant, orally active antagonist of the P2X7 purinergic receptor used in preclinical research to probe P2X7-mediated signaling and inflammatory pathways. It is supplied as a high-purity research compound and is commonly used in both in vitro and in vivo pharmacological studies.