Learn More
Medchemexpress LLC JNJ-47117096 hydroch | 1610536-69-0 | 50MG
SDP
Catalog No. 5000236089
Encompass_Preferred
JNJ-47117096 hydrochloride is potent and selective MELK inhibitor with an IC50 of 23 nM also effectively inhibits Flt3 with an IC50 of 18 nM
Catalog No. 50-002-36089 Supplier Medchemexpress LLC Supplier No. HY1242050MG
JNJ-47117096 hydrochloride is potent and selective MELK inhibitor with an IC50 of 23 nM also effectively inhibits Flt3 with an IC50 of 18 nM