Learn More
Medchemexpress LLC HY-12420 5mg Medchemexpress, JNJ-47117096 hydrochloride CAS:1610536-69-0 Purity:>98%
SDP
Catalog No. 502081038
Encompass_Preferred
Medchemexpress, HY-12420 5mg JNJ-47117096 hydrochloride CAS:1610536-69-0 JNJ-47117096 hydrochloride is potent and selective MELK inhibitor, with an IC50 of 23 nM, also effectively inhibits Flt3, with an IC50 of 18 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1038 Supplier Medchemexpress LLC Supplier No. HY124205MG
Medchemexpress, HY-12420 5mg JNJ-47117096 hydrochloride CAS:1610536-69-0 JNJ-47117096 hydrochloride is potent and selective MELK inhibitor, with an IC50 of 23 nM, also effectively inhibits Flt3, with an IC50 of 18 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.