Learn More
Medchemexpress LLC JNJ-47965567 | 1428327-31-4 | MFCD00283342 | 99.8% | 488.64 g·mol⁻¹ | C28H32N4O2S | 1 ML
SDP
Catalog No. 5000247878
Encompass_Preferred
JNJ-47965567 is a centrally permeable, high-affinity, selective antagonist of the P2X7 purinergic receptor supplied as a 10 mM solution in DMSO for research use. It is used to probe P2X7-mediated signaling in vitro and in vivo and has been applied in rodent CNS pharmacology studies.
  • High affinity and selectivity for P2X7 receptor.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Supplied as a ready-to-use 10 mM solution in DMSO (1 mL).
  • High chemical purity supports reproducible results.
  • Available in multiple solid and solution pack sizes for flexibility.
Catalog No. 50-002-47878 Supplier Medchemexpress LLC Supplier No. HY10141810MM 1ML
JNJ-47965567 is a centrally permeable, high-affinity, selective antagonist of the P2X7 purinergic receptor supplied as a 10 mM solution in DMSO for research use. It is used to probe P2X7-mediated signaling in vitro and in vivo and has been applied in rodent CNS pharmacology studies.
  • High affinity and selectivity for P2X7 receptor.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Supplied as a ready-to-use 10 mM solution in DMSO (1 mL).
  • High chemical purity supports reproducible results.
  • Available in multiple solid and solution pack sizes for flexibility.