Learn More
TARGETMOL CHEMICALS INC JNJ-54175446 1MG
SDP
Catalog No. 502876375
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JNJ-54175446 (JNJ-5446) is a CNS-permeable and selective P2X7 receptor antagonist that attenuates the release of IL-1beta/IL-18 from microglia, and may be used in the study of depression. Purity 99.83%
Catalog No. 50-287-6375 Supplier TARGETMOL CHEMICALS INC Supplier No. T156221MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JNJ-54175446 (JNJ-5446) is a CNS-permeable and selective P2X7 receptor antagonist that attenuates the release of IL-1beta/IL-18 from microglia, and may be used in the study of depression. Purity 99.83%