Learn More
Medchemexpress LLC Jnj-54717793 | 1628843-99-1 | 98.1% | 458.41 | C22H18F4N6O | 5 MG
SDP
Catalog No. 5000222080
Encompass_Preferred
JNJ-54717793 (CAS 1628843-99-1) is a brain-penetrant, orally active, selective, high-affinity orexin-1 receptor (OX1R) antagonist used in neuroscience research. Supplied as an off-white to pink solid, the compound has formula C22H18F4N6O, molecular weight 458.41, and reported purity around 98%.
  • Selective orexin-1 receptor antagonist with high affinity.
  • Marked selectivity versus orexin-2 receptor (lower affinity).
  • Brain penetrant and orally active for in vivo studies.
  • Provided as a powder with recommended cold storage for stability.
  • Supplied in small research quantities suitable for preclinical experiments (5 mg).
Catalog No. 50-002-22080 Supplier Medchemexpress LLC Supplier No. HY1341885MG
JNJ-54717793 (CAS 1628843-99-1) is a brain-penetrant, orally active, selective, high-affinity orexin-1 receptor (OX1R) antagonist used in neuroscience research. Supplied as an off-white to pink solid, the compound has formula C22H18F4N6O, molecular weight 458.41, and reported purity around 98%.
  • Selective orexin-1 receptor antagonist with high affinity.
  • Marked selectivity versus orexin-2 receptor (lower affinity).
  • Brain penetrant and orally active for in vivo studies.
  • Provided as a powder with recommended cold storage for stability.
  • Supplied in small research quantities suitable for preclinical experiments (5 mg).