Learn More
Medchemexpress LLC HY-123857 10mg , JNJ-55308942 CAS:2166558-11-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12385710MG
Encompass_Preferred
HY-123857 10mg JNJ-55308942 CAS: 2166558-11-6 JNJ-55308942 is a high-affinity, selective, brain-penetrant P2X7 functional antagonist (hP2X7: IC50=10 nM, Ki=7.1 nM; rP2X7: IC50=15 nM, Ki=2.9 nM). JNJ-55308942 is orally bioavailable, binds to brain P2X7 and blocks IL-1β release from adult rodent brain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1331