Learn More
Medchemexpress LLC Jnj-55308942 | 2166558-11-6 | 99.9% | 425.32 | C17H12F5N7O | 50MG
SDP
Catalog No. 5000237114
Encompass_Preferred
JNJ-55308942 is a selective, brain-penetrant P2X7 receptor antagonist characterized for preclinical research. It shows low-nanomolar potency against human and rodent P2X7 receptors, reduces IL-1β release in brain tissue, and is supplied as a small research quantity with high reported purity.
  • Selective P2X7 receptor antagonist for target-specific studies
  • Brain-penetrant, suitable for central nervous system research
  • Low-nanomolar potency in human and rodent assays
  • Reduces IL-1β release in brain tissue models
  • High purity (~99.9%) for reliable assay results
  • Provided as a 50 mg research quantity
  • Molecular weight 425.32; formula C17H12F5N7O
Catalog No. 50-002-37114 Supplier Medchemexpress LLC Supplier No. HY12385750MG
JNJ-55308942 is a selective, brain-penetrant P2X7 receptor antagonist characterized for preclinical research. It shows low-nanomolar potency against human and rodent P2X7 receptors, reduces IL-1β release in brain tissue, and is supplied as a small research quantity with high reported purity.
  • Selective P2X7 receptor antagonist for target-specific studies
  • Brain-penetrant, suitable for central nervous system research
  • Low-nanomolar potency in human and rodent assays
  • Reduces IL-1β release in brain tissue models
  • High purity (~99.9%) for reliable assay results
  • Provided as a 50 mg research quantity
  • Molecular weight 425.32; formula C17H12F5N7O