Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JNJ-55308942 | 2166558-11-6 | 99.9% | 425.32 | C17H12F5N7O | 5MG
SDP

Catalog No. 5000235465
Encompass_Preferred

JNJ-55308942 is a selective, brain-penetrant P2X7 receptor antagonist supplied for research use. It shows high affinity for human and rat P2X7 channels and includes solubility and formulation guidance to support both in vitro and in vivo studies.

  • Selective P2X7 antagonist with hP2X7 IC50 = 10 nM and rP2X7 IC50 = 15 nM.
  • Brain-penetrant, suitable for central nervous system studies.
  • Molecular formula C17H12F5N7O; molecular weight 425.32.
  • High purity (≈99.9%).
  • Soluble in DMSO (100 mg/mL); in vivo formulations ≥4 mg/mL reported.
  • Supplied in small milligram pack sizes for research applications.

Catalog No. 50-002-35465 Supplier Medchemexpress LLC Supplier No. HY1238575MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JNJ-55308942 is a selective, brain-penetrant P2X7 receptor antagonist supplied for research use. It shows high affinity for human and rat P2X7 channels and includes solubility and formulation guidance to support both in vitro and in vivo studies.

  • Selective P2X7 antagonist with hP2X7 IC50 = 10 nM and rP2X7 IC50 = 15 nM.
  • Brain-penetrant, suitable for central nervous system studies.
  • Molecular formula C17H12F5N7O; molecular weight 425.32.
  • High purity (≈99.9%).
  • Soluble in DMSO (100 mg/mL); in vivo formulations ≥4 mg/mL reported.
  • Supplied in small milligram pack sizes for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.