Learn More
Medchemexpress LLC JNJ-63576253 | 100MG
SDP
Catalog No. 5000228504
Encompass_Preferred
JNJ-63576253 (TRC-253) is a potent and orally active full antagonist of androgen receptor (AR) with IC50s of 37 and 54 nM for F877L mutant AR and wild-type AR in LNCaP cells JNJ-63576253 can be used for the research of castration-resistant prostate cancer (CRPC)[1]
Catalog No. 50-002-28504 Supplier Medchemexpress LLC Supplier No. HY115282A100MG
JNJ-63576253 (TRC-253) is a potent and orally active full antagonist of androgen receptor (AR) with IC50s of 37 and 54 nM for F877L mutant AR and wild-type AR in LNCaP cells JNJ-63576253 can be used for the research of castration-resistant prostate cancer (CRPC)[1]