Learn More
Medchemexpress LLC HY-101564 1mg , JNJ-64619178 CAS:2086772-26-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015641MG
Encompass_Preferred
HY-101564 1mg JNJ-64619178 CAS: 2086772-26-9 JNJ-64619178 (Onametostat) is a selective, orally active and pseudo-irreversible protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 0.14 nM. JNJ-64619178 has potent activity In lung cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9290