Learn More
Selleck Chemical LLC JNJ-64619178 1mg 2086772-26-9
SDP
Supplier:  Selleck Chemical LLC S86241MG
Encompass_Preferred
JNJ-64619178 is a PRMT5 inhibitor with high selectivity and potency (subnanomolar range, PRMT5-MEP-50 IC50=0.14 nM) under different in vitro and cellular conditions, paired with favorable pharmacokinetics and safety properties. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1423276