Learn More
Medchemexpress LLC HY-139399 10mg , JNJ-67856633 CAS:2230273-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13939910MG
Encompass_Preferred
HY-139399 10mg JNJ-67856633 CAS: 2230273-76-2 JNJ-67856633 is an orally active, first-in-class, potent, selective and allosteric MALT1 protease inhibitor. JNJ-67856633 in some cases lead to tumor stasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0829