Learn More
Cayman Chemical JNJ-7706621, Purity: ≥98%, CAS: 443797-96-4, 10 mg
SDP
Catalog No. 502980028
Encompass_Preferred
A dual inhibitor of Cdks and Aurora kinases; inhibits Cdk1/cyclin B, Cdk2/cyclin A, Cdk2/cyclin E, Cdk3/cyclin E, Cdk4/cyclin D1, Cdk6/cyclin D1, Aurora A, and Aurora B in vitro (IC50s = 9, 4, 3, 58, 253, 175, 11, and 15 nM, respectively); blocks the growth of cancer cells in vitro and in vivo
Catalog No. 50-298-0028 Supplier Cayman Chemical Supplier No. 1849410 MG
A dual inhibitor of Cdks and Aurora kinases; inhibits Cdk1/cyclin B, Cdk2/cyclin A, Cdk2/cyclin E, Cdk3/cyclin E, Cdk4/cyclin D1, Cdk6/cyclin D1, Aurora A, and Aurora B in vitro (IC50s = 9, 4, 3, 58, 253, 175, 11, and 15 nM, respectively); blocks the growth of cancer cells in vitro and in vivo