Learn More
Medchemexpress LLC HY-10329 2mg Medchemexpress, JNJ-7706621 CAS:443797-96-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103292MG
Encompass_Preferred
Medchemexpress, HY-10329 2mg JNJ-7706621 CAS:443797-96-4 JNJ-7706621 is a potent aurora kinase inhibitor, and also inhibits CDK1 and CDK2, with IC50s of 9, 3, 11, and 15 nM for CDK1, CDK2, Aurora-A and Aurora-B, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8068