Learn More
Selleck Chemical LLC JNJ-7706621 5mg 443797-96-4
SDP
Catalog No. 508801
Encompass_Preferred
JNJ-7706621 is pan-CDK inhibitor with the highest potency on CDK1/2 with IC50 of 9 nM/4 nM and showing >6-fold selectivity for CDK1/2 than CDK3/4/6 in cell-free assays. It also potently inhibits Aurora A/B and has no activity on Plk1 and Wee1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-880-1 Supplier Selleck Chemical LLC Supplier No. S12495MG
JNJ-7706621 is pan-CDK inhibitor with the highest potency on CDK1/2 with IC50 of 9 nM/4 nM and showing >6-fold selectivity for CDK1/2 than CDK3/4/6 in cell-free assays. It also potently inhibits Aurora A/B and has no activity on Plk1 and Wee1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.