Learn More
Medchemexpress LLC Jnj-8003 | 3118087-68-3 | 99.8% | 576.51 | 1 ML
SDP
Catalog No. 5000359463
Encompass_Preferred
JNJ-8003 is a potent and orally active non-nucleoside RSV polymerase inhibitor. It targets the L protein polymerase complex of RSV, blocking the transcription and replication of the viral genome by inhibiting RNA-dependent RNA polymerase (RdRp) activity. It demonstrates subnanomolar activity in vitro and prominent efficacy in mice and neonatal lamb models, making it suitable for respiratory syncytial virus (RSV) research.
  • Potent and orally active non-nucleoside RSV polymerase inhibitor
  • Targets the L protein polymerase complex of RSV
  • Blocks transcription and replication of viral genome
  • Inhibits RNA-dependent RNA polymerase (RdRp) activity
  • Displays subnanomolar activity in vitro
  • Prominent efficacy in mice and neonatal lamb models
  • Used for the study of respiratory syncytial virus (RSV)
Catalog No. 50-003-59463 Supplier Medchemexpress LLC Supplier No. HY14964810MM/1ML
JNJ-8003 is a potent and orally active non-nucleoside RSV polymerase inhibitor. It targets the L protein polymerase complex of RSV, blocking the transcription and replication of the viral genome by inhibiting RNA-dependent RNA polymerase (RdRp) activity. It demonstrates subnanomolar activity in vitro and prominent efficacy in mice and neonatal lamb models, making it suitable for respiratory syncytial virus (RSV) research.
  • Potent and orally active non-nucleoside RSV polymerase inhibitor
  • Targets the L protein polymerase complex of RSV
  • Blocks transcription and replication of viral genome
  • Inhibits RNA-dependent RNA polymerase (RdRp) activity
  • Displays subnanomolar activity in vitro
  • Prominent efficacy in mice and neonatal lamb models
  • Used for the study of respiratory syncytial virus (RSV)