Learn More
TARGETMOL CHEMICALS INC JNJ-A07 1MG
SDP
Catalog No. 502879820
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (+)-JNJ-A07 is a pan-serotypic dengue virus inhibitor that targets NS3-NS4B interaction with high potency. (+)-JNJ-A07 has antiviral activity and can be used to study dengue virus infections. Purity 99.93%
Catalog No. 50-287-9820 Supplier TARGETMOL CHEMICALS INC Supplier No. T640921MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (+)-JNJ-A07 is a pan-serotypic dengue virus inhibitor that targets NS3-NS4B interaction with high potency. (+)-JNJ-A07 has antiviral activity and can be used to study dengue virus infections. Purity 99.93%