Learn More
TARGETMOL CHEMICALS INC JNJ-A07 1MG
Supplier: TARGETMOL CHEMICALS INC T640911MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (-)-JNJ-A07 is a selective and potent dengue virus (DENV) inhibitor with an EC50 of 31 nM, exhibiting significant antiviral activity for the study of dengue virus infections. Purity 99.75%