Learn More
Medchemexpress LLC (-)-JNJ-A07 | 2135640-92-3 | 100.0% | 578.96 | 100 MG
SDP
Catalog No. 5000349274
Encompass_Preferred
(-)-JNJ-A07 is a potent and selective DENV inhibitor with an EC50 value of 31 nM. It is for research use only and not sold to patients.
  • Potent and selective DENV inhibitor
  • EC50 value of 31 nM
  • For research use only
  • Not sold to patients
Catalog No. 50-003-49274 Supplier Medchemexpress LLC Supplier No. HY139602C100MG
(-)-JNJ-A07 is a potent and selective DENV inhibitor with an EC50 value of 31 nM. It is for research use only and not sold to patients.
  • Potent and selective DENV inhibitor
  • EC50 value of 31 nM
  • For research use only
  • Not sold to patients